missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 20mL PET Liquid Scintillation Vials, With Caps Attached

For optimal performance in the laboratory. Wheaton™ 20mL PET Liquid Scintillation Vials, With Caps Attached offer low permeability to solvents and minimal background counts.

Brand:  DWK Life Sciences 986731

260.37 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11758929

  • £333.00 / Pack of 500
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Clarity of glass with the safety of plastic
  • Vial can be incinerated; no harmful gas is generated
  • Vials offer low permeability to solvents and minimal background counts

 

Order Info

White. Packaged in utility trays with each partitioned tray holding 100 vials.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Metal Foil/Pulp
White
PET
Offer low permeability to solvents and minimal background counts, vials can be safely incinerated, no harmful gas is generated
Tray
Polypropylene
61 mm
1.06 in.
27 mm
20 mL
2.4 in.
0.67 oz.
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ 20mL PET Liquid Scintillation Vials, With Caps Attached > PP Cap, Metal Foil/Pulp Liner, Screw Caps Attached to Vials; 500/Cs.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.