missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 25mm Filtration Assemblies and Accessories

Choose from a wide range of filtration assemblies and accessories to suit your application needs

Brand:  DWK Life Sciences 635229

48.56 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12487121

  • £65.80 / Each
Estimated Shipment: 03-10-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP Type I, ASTM E 438 Type I, Class A
25mm filtration assembly
Analysis of particulate or microbiological contamination
1 Case
125 mL
0.25 in. hose
Borosilicate Glass
Yes
Filtration Flask
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ 25mm Filtration Assemblies and Accessories > Filter Flask, 125mL; Graduated; No. 5 Stopper Joint

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.