missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 25mm Filtration Assembly With No. 5 Stopper Connection With Stainless Steel Support

Designed to handle small volumes of liquids for analysis of particulate or microbiological contamination

Brand:  DWK Life Sciences 419327

249.91 GBP valid until 2025-12-31
Use promo code "24293" to get your promotional price.


 View more versions of this product

Product Code. 11791893

  • £276.00 / Each
Estimated Shipment: 27-08-2025
to see stock.
Explore more special offers
Add to basket

Includes:
  • 316 stainless steel support with 120 mesh screen
  • PTFE support screen gasket
  • Glass support base
  • 15mL graduated funnel
  • Anodized aluminum clamp
  • No. 5 silicone stopper
This item is not returnable. View return policy

Description

Description

  • For filtration of viscous or proteinaceous solutions to produce ultra clean filtrate
  • Glass components manufactured from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements

TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP Type I, ASTM E 438 Type I, Class A
Borosilicate Glass
One 316 stainless steel support with 120 mesh screen, PTFE support screen gasket, glass support base, 15mL graduated funnel, anodized aluminum clamp, one No. 5 silicone stopper
Filtration Assembly
25mm filtration assembly
Filtering viscous or proteinaceous solutions, to produce ultraclean filtrate
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ 25mm Filtration Assembly With No. 5 Stopper Connection With Stainless Steel Support > Filtration Assembly; 25mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.