Learn More
DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
£231.00 - £1635.00
Specifications
Material Borosilicate Glass
View More Specs
Products 15
Product Code Brand Capacity Color Height (English) Price Quantity & Availability  
Product Code Brand Capacity Color Height (English) Price Quantity & Availability  
11591165
View accessory products View Documents
DWK Life Sciences
W223712
0.1 oz., 2 mL Clear 1.41 in.
£231.00
Pack of 288
Estimated Shipment: 09-06-2025
to see stock.
Add to basket
10520853
View accessory products View Documents
DWK Life Sciences
223738
0.17 oz., 5mL Clear 1.85 in.
£255.00
Pack of 288
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
10424292
View accessory products View Documents
DWK Life Sciences
223760
0.17 oz., 5 mL Amber 1.85 in.
£461.00
Pack of 288
Estimated Shipment: 11-08-2025
to see stock.
Add to basket
10768134
View accessory products View Documents
DWK Life Sciences
223739
0.34 oz., 10 mL Clear 2.12 in.
£424.00
Pack of 288
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
10434292
View accessory products View Documents
DWK Life Sciences
223761
0.34 oz., 10 mL Amber 2.12 in.
£710.00
Pack of 288
Estimated Shipment: 14-07-2025
to see stock.
Add to basket
10453712
View accessory products View Documents
DWK Life Sciences
223742
0.7 oz., 20 mL Clear 2.28 in.
£426.00
Pack of 288
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
10600173
View accessory products View Documents
DWK Life Sciences
223762
0.7 oz., 20 mL Amber 2.28 in.
£1635.00
Pack of 288
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
10071392
View accessory products View Documents
DWK Life Sciences
223763
1.01 oz., 30 mL Amber 2.44 in.
£886.00
Pack of 288
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
10632973
View accessory products View Documents
DWK Life Sciences
223743
1.01 oz., 30 mL Clear 2.48 in.
£524.00
Pack of 288
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
10464672
View accessory products View Documents
DWK Life Sciences
223764
1.7 oz., 50 mL Amber 2.87 in.
£1210.00
Pack of 288
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
10642973
View accessory products View Documents
DWK Life Sciences
223745
1.7 oz., 50 mL Clear 2.87 in.
£911.00
Pack of 288
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
10235262
View Documents
DWK Life Sciences
223746
2.03 oz., 60 mL Clear 3.58 in.
£371.00
Pack of 144
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
10011422
View accessory products View Documents
DWK Life Sciences
223766
3.38 oz., 100 mL Amber 3.74 in.
£870.00
Pack of 144
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
10021362
View accessory products View Documents
DWK Life Sciences
223747
3.38 oz., 100 mL Clear 3.74 in.
£1545.00
Pack of 144
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
10071382
View accessory products View Documents
DWK Life Sciences
223748
4.23 oz., 125 mL Clear 4.21 in.
£715.00
Pack of 144
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
Description
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage
Compliance
Meet requirements for Class A of the United States Pharmacopeia for Type I glass.
Specifications
Specifications
Borosilicate Glass