Learn More
Waters Corporation XBridge BEH HILIC XP Column
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds.
Brand: Waters Corporation 176002583

Description
- These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
- More rugged than silica based HILIC phases, the BEH particle's chemical stability results in long column lifetimes.
- Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Specifications
3 mm | |
18000 psi (1240 Bar) | |
2.5 μm | |
3/Pk. | |
No |
50 mm | |
Spherical | |
130 Å | |
L3 | |
eXtended Performance (XP) Column |
Product Suggestions
Customers who viewed this item also viewed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.