Learn More
Waters Corporation XBridge C18 Column
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1 to 12).
£832.50 - £1170.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
10309612
|
Waters Corporation
186003117 |
4.6 mm | 250 mm | 5 μm |
£1132.50
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
13222960
|
Waters Corporation
186003020 |
2.1 mm | 30 mm | 3.5 μm |
£907.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
10729006
|
Waters Corporation
186003021 |
2.1 mm | 50 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
10614596
|
Waters Corporation
186003022 |
2.1 mm | 100 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
13262960
|
Waters Corporation
186003025 |
3 mm | 30 mm | 3.5 μm |
£907.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13272960
|
Waters Corporation
186003026 |
3 mm | 50 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
10015683
|
Waters Corporation
186003027 |
3 mm | 100 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||
10136503
|
Waters Corporation
186003028 |
3 mm | 150 mm | 3.5 μm |
£1072.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13292960
|
Waters Corporation
186003030 |
4.6 mm | 30 mm | 3.5 μm |
£907.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
10614776
|
Waters Corporation
186003031 |
4.6 mm | 50 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13212970
|
Waters Corporation
186003032 |
4.6 mm | 75 mm | 3.5 μm |
£952.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
11916938
|
Waters Corporation
186003033 |
4.6 mm | 100 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||
10779186
|
Waters Corporation
186003034 |
4.6 mm | 150 mm | 3.5 μm |
£1072.50
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
13203020
|
Waters Corporation
186003107 |
2.1 mm | 20 mm | 5 μm |
£832.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13213020
|
Waters Corporation
186003108 |
2.1 mm | 50 mm | 5 μm |
£900.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13253020
|
Waters Corporation
186003113 |
4.6 mm | 50 mm | 5 μm |
£900.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13263020
|
Waters Corporation
186003114 |
4.6 mm | 75 mm | 5 μm |
£937.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13273020
|
Waters Corporation
186003115 |
4.6 mm | 100 mm | 5 μm |
£967.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
15315255
|
Waters Corporation
186003116 |
4.6 mm | 150 mm | 5 μm |
£1035.00
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
13223030
|
Waters Corporation
186003126 |
1 mm | 50 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13233030
|
Waters Corporation
186003128 |
1 mm | 150 mm | 3.5 μm |
£1072.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13273030
|
Waters Corporation
186003132 |
3 mm | 100 mm | 5 μm |
£967.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
10537407
|
Waters Corporation
186003109 |
2.1 mm | 100 mm | 5 μm |
£967.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
11926938
|
Waters Corporation
186003110 |
2.1 mm | 150 mm | 5 μm |
£1035.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
10034645
|
Waters Corporation
186003111 |
3 mm | 30 mm | 5 μm |
£877.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13243020
|
Waters Corporation
186003112 |
3 mm | 150 mm | 5 μm |
£1035.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
10004853
|
Waters Corporation
186003127 |
1 mm | 100 mm | 5 μm |
£1005.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13243030
|
Waters Corporation
186003129 |
2.1 mm | 30 mm | 5 μm |
£877.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13263030
|
Waters Corporation
186003131 |
3 mm | 50 mm | 5 μm |
£900.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13283030
|
Waters Corporation
186003133 |
3 mm | 250 mm | 5 μm |
£1132.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13203040
|
Waters Corporation
186003135 |
4.6 mm | 30 mm | 5 μm |
£877.50
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
13233270
|
Waters Corporation
186003943 |
4.6 mm | 250 mm | 3.5 μm |
£1170.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
Description
- The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.
Specifications
6000 psi (415 Bar) | |
130 Å | |
L1 | |
Column |
Spherical | |
1/Ea. | |
Yes |