missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ XL 184

Potent VEGFR inhibitor; also inhibits other RTKs

£206.00 - £781.00

Specifications

Purity >98%
Gene Alias Cabozantinib, BMS 907351
Product Type XL 184
Molecular Weight (g/mol) 501.51
Inhibitors XL 184
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18716229
View Documents
Tocris Bioscience™
5422/10
10 mg
£206.00
10mg
Please to purchase this item. Need a web account? Register with us today!
18734909
View Documents
Tocris Bioscience™
5422/50
50 mg
£781.00
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent VEGFR inhibitor (IC50 = 0.035 nM); also inhibits c-Met, KIT, RET, FLT4, AXL, FLT3, FLT1 and Tie2 (IC50 values are 1.3, 4.6, 5.2, 6, 7, 11.3, 12 and 14.3 nM, respectively). Induces intratumoral hypoxia and apoptosis. Reduces tumor invasion and metastasis in vivo. Antiangiogenic.
Specifications

Specifications

>98%
XL 184
XL 184
Cabozantinib, BMS 907351
501.51
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.