missing translation for 'onlineSavingsMsg'
Learn More
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals Product Code.: 12632916

Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals

Product Code. 12632916
2.5 L, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 12632916

Brand: Thermo Scientific Acros 447240025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -34.0°C
Density 0.8650g/mL
Boiling Point 136.0°C to 140.0°C
Quantity 2.5 L
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4940 to 1.4980
Assay Percent Range 0.4% max. Toluene (GC)
Specific Gravity 0.865
Linear Formula C6H4(CH3)2
Viscosity 0.6 mPa.s (20°C)
Merck Index 15,10278
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,methylene chloride,acetone and ether
Formula Weight 106.17
Percent Purity 99%
Grade Biochemical/Histological
Chemical Name or Material Xylenes
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467707 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number447240010,447240025
Lot NumberA0460582 Certificate TypeCertificate of Analysis Date27/02/2024 Catalog Number447240010,447240025
Lot NumberA0450918 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number447240010,447240025
Lot NumberA0415914 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number447240010,447240025
Lot NumberA0446153 Certificate TypeCertificate of Analysis Date28/09/2022 Catalog Number447240010,447240025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylenes
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H304-May be fatal if swallowed and enters airways.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.