missing translation for 'onlineSavingsMsg'
Learn More
Xylenes, mixed, 97+%, Thermo Scientific Chemicals Product Code.: 11318036

Xylenes, mixed, 97+%, Thermo Scientific Chemicals

Product Code. 11318036
500 mL
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11318036

Brand: Thermo Scientific Alfa Aesar L13317.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Xylene is used as a solvent in pharmaceutical, printing, rubber and leather industries. It is an important component of ink, rubber and adhesives as well as thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xylene is used as a solvent in pharmaceutical, printing, rubber and leather industries. It is an important component of ink, rubber and adhesives as well as thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.

Solubility
Miscible with aromatic hydrocarbons. Immiscible with water.

Notes
Contains a mixture of the ortho, meta, and para isomers of xylene. May also contain ethylbenzene. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
MDL Number MFCD00077264
Synonym Dimethylbenzenes + ethylbenzene

Specifications

Melting Point <-25°C
Density 0.86
Boiling Point 137°C to 144°C
Quantity 500 mL
Flash Point 29°C (84°F)
Odor Aromatic
Refractive Index 1.497
UN Number UN1307
Beilstein 1901563
Merck Index 14,10081
Solubility Information Miscible with aromatic hydrocarbons. Immiscible with water.
Formula Weight 106.17
Percent Purity ≥97%
Chemical Name or Material Xylenes, mixed
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2420730 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberL13317.0F,L13317.AP,L13317.AU
Lot Number10224028 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL13317.0F,L13317.AP,L13317.AU
Lot Number10237726 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog NumberL13317.0F,L13317.AP,L13317.AU
Lot Number10238541 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL13317.0F,L13317.AP,L13317.AU
Lot Number10220799 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog NumberL13317.0F,L13317.AP,L13317.AU

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Xylenes, mixed, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.