missing translation for 'onlineSavingsMsg'
Learn More
Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals Product Code.: 10453931

Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals

Product Code. 10453931
500 mL
Click to view available options
Quantity:
500 mL
2.5 L
25 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10453931

Brand: Thermo Scientific Acros 422685000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7, 100-41-4
Molecular Formula C8H10
MDL Number MFCD00077264
Synonym Dimethylbenzene

Specifications

Melting Point -34.0°C
Density 0.8650g/mL
Boiling Point 136.0°C to 140.0°C
Quantity 500 mL
Flash Point 23°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4940 to 1.4980
Assay Percent Range 99%
Residue after Evaporation 0.002% max.
Specific Gravity 0.865
Linear Formula C6H4(CH3)2
Vapor Pressure Vapor Pressure: 8mbar at 20°C
Viscosity 0.6 mPa.s (20°C)
Beilstein 05, 360
Water 0.05% max. (Coulometric)
Merck Index 15, 10278
Solubility Information Solubility in water: <0.1g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride, acetone and ether
Formula Weight 106.17
Percent Purity 99%
Grade ACS Reagent
Chemical Name or Material Xylenes
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467581 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number422680025,422680250,422685000
Lot NumberB0546495A Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number422680025,422680250,422685000
Lot NumberA0461056 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number422680025,422680250,422685000
Lot NumberB0542026A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number422680025,422680250,422685000
Lot NumberB0541298A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number422680025,422680250,422685000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.