missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Yeast Lytic Enzyme, Arthrobacter luteus
Hydrolyzes linear glucose polymers with beta-1,3-linkages
£306.00 - £658.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15445459
|
Thermo Scientific Chemicals
J63195.MD |
250 mg |
£306.00
250mg |
In Stock
Log in to see stock. |
|||||
15455459
|
Thermo Scientific Chemicals
J63195.ME |
500 mg |
£522.00
500mg |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
15435459
|
Thermo Scientific Chemicals
J63195.03 |
1 g |
£658.00
1g |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
Description
Yeast Lytic Enzyme is used to hydrolyzes linear glucose polymers with beta-1,3-linkages, used for enzymatic digestion of yeast glycan coats and for spheroplast formation, used to digest the cell wall of active yeast cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
37340-57-1 | |
Lyticase; Zymolyase 20T |
Specifications
Off-white | |
37340-57-1 | |
2°C to 8°C | |
Hydrolyzes linear glucose polymers with beta-1,3-linkages |
Cell Lysis Enzyme | |
Lyticase; Zymolyase 20T | |
Lyophilized |
Safety and Handling
Recommended Storage : Keep cold
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Yeast Lytic Enzyme, Arthrobacter luteus
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only