missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Z-Leu-Val-Gly Diazomethyl Ketone Product Code.: 15496549

Thermo Scientific Chemicals Z-Leu-Val-Gly Diazomethyl Ketone

Product Code. 15496549
25 mg
Click to view available options
Quantity:
25 mg
This item is not returnable. View return policy

Product Code. 15496549

Brand: Thermo Scientific Chemicals J64731.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119670-30-3 | C22H31N5O5, C22H31N5O5 | 445.52 g/mol

Mimics part of the human cysteine proteinase-binding center; an irreversible inhibitor of human cysteine proteinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Mimics part of the human cysteine proteinase-binding center
  • An irreversible inhibitor of human cysteine proteinase
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Mimics part of the human cysteine proteinase-binding center; an irreversible inhibitor of human cysteine proteinase
CAS 119670-30-3
Molecular Formula C22H31N5O5, C22H31N5O5
Formula Weight 445.52g/mol
Synonym C-3455
Product Type Signal Transduction Reagent
Form Powder
InChI Key ZLDBPOCJCXPVND-OALUTQOASA-N
SMILES CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)NC(=O)[C@@H](NCC(=O)C=[N+]=[N-])C(C)C
IUPAC Name benzyl N-[(1S)-1-{[(2S)-2-[(3-diazo-2-oxopropyl)amino]-3-methylbutanoyl]carbamoyl}-3-methylbutyl]carbamate
Molecular Weight (g/mol) 445.52
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM10J013 Certificate TypeCertificate of Analysis Date19/01/2023 Catalog NumberJ64731.MB
Lot NumberQ30G023 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ64731.MB
Lot NumberQ30G015 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ64731.MB
Lot NumberZ02B016 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ64731.MB

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Z-Leu-Val-Gly Diazomethyl Ketone >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.