Learn More
Zinc sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Zinc sputtering target is used in the manufacture of paints, bleaches and to make other chemicals. It is also used in medical research. Pure zinc is used as an electrode in a Daniel cell and also in dry batteries. It is used as a galvanic coating on steel to prevent corrosion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 1 Ea. |
CAS | 7440-66-6 |
Odor | Odorless |
Molecular Formula | Zn |
MDL Number | MFCD00011291 |
Synonym | dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust |
Solubility Information | Insoluble in water. |
InChI Key | HCHKCACWOHOZIP-UHFFFAOYSA-N |
SMILES | [Zn] |
IUPAC Name | zinc |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
J25M22 | Certificate of Analysis | 07/12/2021 | 041656.KS |
D01P19 | Certificate of Analysis | 07/12/2021 | 041656.KS |
K06S048 | Certificate of Analysis | 07/12/2021 | 041656.KS |
M29F053 | Certificate of Analysis | 07/12/2021 | 041656.KS |
C02X065 | Certificate of Analysis | 07/12/2021 | 041656.KS |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.