Learn More
Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
£36.78 - £406.02
Chemical Identifiers
CAS | 7446-20-0 |
---|---|
Molecular Formula | H14O11SZn |
Molecular Weight (g/mol) | 287.54 |
MDL Number | MFCD00149894 |
InChI Key | RZLVQBNCHSJZPX-UHFFFAOYSA-L |
Synonym | zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o |
PubChem CID | 62640 |
ChEBI | CHEBI:32312 |
IUPAC Name | zinc;sulfate;heptahydrate |
SMILES | O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10703211
|
Thermo Scientific Acros
205982500 |
250 g | Plastic bottle |
£36.78
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666252
|
Thermo Scientific Acros
205980010 |
1 kg | Plastic bottle |
£106.53
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10267163
|
Thermo Scientific Acros
205980050 |
5 kg | Plastic bottle |
£406.02
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7446-20-0 | |
287.54 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
62640 | |
zinc;sulfate;heptahydrate |
H14O11SZn | |
MFCD00149894 | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
CHEBI:32312 | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Specifications
100.0°C | |
White | |
Crystalline Powder and/or Crystals | |
H14O11SZn | |
MFCD00149894 | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
zinc;sulfate;heptahydrate | |
62640 | |
287.53 | |
Analytical |
7446-20-0 | |
4.4 to 6 (5% soln. at 20°C) | |
99.4% min. (Complexometry) | |
ZnSO4·7H2O | |
15, 10359 | |
Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol, insoluble in alcohol | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
287.54 | |
CHEBI:32312 | |
99.5% | |
Zinc sulfate heptahydrate |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Zinc sulfate heptahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.