missing translation for 'onlineSavingsMsg'
Learn More
Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals Product Code.: 11469463

Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11469463
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11469463

Brand: Thermo Scientific Alfa Aesar A12342.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

Solubility
Soluble in water and alcohol.

Notes
Hygroscopic, moisture sensitive and incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13520-92-8
Molecular Formula Cl2H16O9Zr
Molecular Weight (g/mol) 322.24
MDL Number MFCD00149898
InChI Key SDFOCNSOSARAKC-UHFFFAOYSA-L
Synonym zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402Show MoreShow Less
PubChem CID 159678
IUPAC Name oxozirconium;octahydrate;dihydrochloride
SMILES O.Cl[Zr](Cl)=O

Specifications

Melting Point 150°C (decomposition)
Quantity 100 g
UN Number UN3260
Merck Index 14,10184
Solubility Information Soluble in water and alcohol.
Formula Weight 322.25 (178.13 Anhydrous)
Percent Purity 98%
Odor Acidic
Sensitivity Hygroscopic
Density 1.91
Chemical Name or Material Zirconium dichloride oxide octahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252827 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog NumberA12342.22,A12342.0E,A12342.36
Lot Number10247126 Certificate TypeCertificate of Analysis Date27/06/2023 Catalog NumberA12342.22,A12342.0E,A12342,A12342.36
Lot Number10242004 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog NumberA12342.22,A12342.0E,A12342.36
Lot Number10241657 Certificate TypeCertificate of Analysis Date05/07/2022 Catalog NumberA12342.22,A12342.0E,A12342.36
Lot Number10239523 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA12342.22,A12342.0E,A12342.36
compliance-icons
Product Identifier
  • Zirconium dichloride oxide octahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Zirconium dichlorideoxide octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.