Learn More
Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals
Description
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.
Solubility
Soluble in water and alcohol.
Notes
Hygroscopic, moisture sensitive and incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 150°C (decomposition) |
Quantity | 500 g |
UN Number | UN3260 |
Merck Index | 14,10184 |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 322.25 (178.13 Anhydrous) |
Percent Purity | 98% |
Odor | Acidic |
Sensitivity | Hygroscopic |
Density | 1.91 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252827 | Certificate of Analysis | 22/10/2024 | A12342.22,A12342.0E,A12342.36 |
10247126 | Certificate of Analysis | 27/06/2023 | A12342.22,A12342.0E,A12342,A12342.36 |
10242004 | Certificate of Analysis | 29/07/2022 | A12342.22,A12342.0E,A12342.36 |
10241657 | Certificate of Analysis | 05/07/2022 | A12342.22,A12342.0E,A12342.36 |
10239523 | Certificate of Analysis | 12/04/2022 | A12342.22,A12342.0E,A12342.36 |
Safety and Handling

- Zirconium dichloride oxide octahydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Zirconium dichlorideoxide octahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.