missing translation for 'onlineSavingsMsg'
Learn More
Zirconyl chloride octahydrate, 98%, Thermo Scientific Chemicals Product Code.: 10125693

Zirconyl chloride octahydrate, 98%, Thermo Scientific Chemicals

Product Code. 10125693
500 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10125693

Brand: Thermo Scientific Acros 208375000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13520-92-8
Molecular Formula Cl2H16O9Zr
Molecular Weight (g/mol) 322.24
MDL Number MFCD00149898
InChI Key SDFOCNSOSARAKC-UHFFFAOYSA-L
Synonym zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402Show MoreShow Less
PubChem CID 159678
IUPAC Name oxozirconium;octahydrate;dihydrochloride
SMILES O.Cl[Zr](Cl)=O

Specifications

Color White to Yellow
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 97.5 to 102.5% (Argentometry)
Linear Formula ZrOCl2·8H2O
Merck Index 15, 10384
Solubility Information Solubility in water: soluble in water. Other solubilities: freely soluble in alcohol,insoluble in other organic solvents
Formula Weight 322.25
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 1.91
Density 1.9100g/mL
Chemical Name or Material Zirconyl chloride octahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467414 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number208375000,208371000,208370050
Lot NumberA0460939 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number208375000,208371000,208370050
Lot NumberA0453895 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number208375000,208371000,208370050
Lot NumberA0448682 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number208375000,208371000,208370050
Lot NumberA0399044 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number208375000,208371000,208370050
compliance-icons
Product Identifier
  • Zirconyl chloride octahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Zirconyl chloride octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconyl chloride octahydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.