missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconyl chloride octahydrate, 98%, Thermo Scientific Chemicals
£20.60 - £237.18
Chemical Identifiers
CAS | 13520-92-8 |
---|---|
Molecular Formula | Cl2H18O9Zr |
Molecular Weight (g/mol) | 324.26 |
MDL Number | MFCD00149898 |
InChI Key | FUBACIUATZGHAC-UHFFFAOYSA-N |
Synonym | zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402 |
PubChem CID | 159678 |
IUPAC Name | oxozirconium;octahydrate;dihydrochloride |
SMILES | O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10656782
|
Thermo Scientific Acros
208370050 |
5 g | Glass bottle |
£20.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10676782
|
Thermo Scientific Acros
208371000 |
100 g | Glass bottle |
£77.79
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10125693
|
Thermo Scientific Acros
208375000 |
500 g | Glass bottle |
£237.18
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13520-92-8 | |
324.26 | |
FUBACIUATZGHAC-UHFFFAOYSA-N | |
159678 | |
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr] |
Cl2H18O9Zr | |
MFCD00149898 | |
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402 | |
oxozirconium;octahydrate;dihydrochloride |
Specifications
13520-92-8 | |
Crystalline Powder | |
Cl2H18O9Zr | |
MFCD00149898 | |
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402 | |
FUBACIUATZGHAC-UHFFFAOYSA-N | |
oxozirconium;octahydrate;dihydrochloride | |
159678 | |
98% | |
1.9100g/mL |
White to Yellow | |
97.5 to 102.5% (Argentometry) | |
ZrOCl2·8H2O | |
15, 10384 | |
(10% in water) Clear to slightly hazy solution | |
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr] | |
324.26 | |
322.25 | |
1.91 | |
Zirconyl chloride octahydrate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Zirconyl chloride octahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Zirconyl chloride octahydrate
RUO – Research Use Only