missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zoniporide dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Sodium-hydrogen exchanger isoform 1 (NHE1) inhibitor that displays selectivity over other NHE isoforms (Ki values are 14, 2200 and 220000 nM for human NHE1, human NHE2 and rat NHE3 respectively). Inhibits NHE1 dependent 22Na+ uptake in vitro (IC50 = 14 nM) and provides cardioprotection from myocardial ischemic injury in vivo (EC50 = 0.25 nM). Also inhibits MMP2/9 activity and invasion in breast cancer cells.

Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 393.27 |
Percent Purity | >99% |
Chemical Name or Material | Zoniporide dihydrochloride |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zoniporide dihydrochloride, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction