missing translation for 'onlineSavingsMsg'
Learn More
1,2-Ethylenediphosphonic acid, 97%, Thermo Scientific Chemicals Product Code.: 11337605

1,2-Ethylenediphosphonic acid, 97%, Thermo Scientific Chemicals

Product Code. 11337605
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11337605

Brand: Thermo Scientific Alfa Aesar L05828.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6145-31-9
Molecular Formula C2H8O6P2
Molecular Weight (g/mol) 190.028
MDL Number MFCD00013958
InChI Key XYJLPCAKKYOLGU-UHFFFAOYSA-N
Synonym 1,2-ethylenediphosphonic acid, 1,2-ethanediphosphonic acid, phosphonic acid, ethylenedi, phosphonic acid, 1,2-ethanediylbis, 1,2-ethanebisphosphonic acid, 1,2-ethanediylbisphosphonic acid, ethylenediphosphonic acid, phosphonic acid, ethylenedi-6ci,7ci,8ci, ethane-1,2-diylbis phosphonicacid, ethylenebisphosphonic acidShow MoreShow Less
PubChem CID 80247
IUPAC Name 2-phosphonoethylphosphonic acid
SMILES C(CP(=O)(O)O)P(=O)(O)O

Specifications

Melting Point 217°C to 221°C
Quantity 25 g
UN Number UN3261
Beilstein 1780250
Formula Weight 190.03
Percent Purity 97%
Chemical Name or Material 1,2-Ethylenediphosphonic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10212828 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL05828.14,L05828.06
Lot Number10136733 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL05828.14,L05828.06
Lot Number10236247 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL05828.14,L05828.06
Lot Number10196270 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL05828.14,L05828.06
Lot NumberGBFA013574 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL05828.14,L05828.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Ethylenediphosphonic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.