missing translation for 'onlineSavingsMsg'
Learn More
1-(3-Aminopropyl)pyrrolidine, 97%, Thermo Scientific Chemicals Product Code.: 11321697

1-(3-Aminopropyl)pyrrolidine, 97%, Thermo Scientific Chemicals

Product Code. 11321697
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11321697

Brand: Thermo Scientific Alfa Aesar L04739.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-(3-Aminopropyl)pyrrolidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It can be used to make other chemicals It can react with 7-chloro-6-(2-chloro-ethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine to get 5-methyl-8-(3-pyrrolidin-1-yl-propyl)-7,8-dihydro-6H-1,3,4,8,8b-pentaaza-as-indacene . It can also react with 2,4-diphenyl-cyclobutane-1,3-dicarbonyl dichloride to obtain 2,4-diphenyl-cyclobutane-1,3-dicarboxylic acid bis-[(3-pyrrolidin-1-yl-propyl)-amide] .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)pyrrolidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It can be used to make other chemicals It can react with 7-chloro-6-(2-chloro-ethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine to get 5-methyl-8-(3-pyrrolidin-1-yl-propyl)-7,8-dihydro-6H-1,3,4,8,8b-pentaaza-as-indacene . It can also react with 2,4-diphenyl-cyclobutane-1,3-dicarbonyl dichloride to obtain 2,4-diphenyl-cyclobutane-1,3-dicarboxylic acid bis-[(3-pyrrolidin-1-yl-propyl)-amide] .

Solubility
Soluble in water, soluble in chloroform

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23159-07-1
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00014100
InChI Key VPBWZBGZWHDNKL-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-yl propan-1-amine, 1-3-aminopropyl pyrrolidine, 1-3-aminopropyl-pyrrolidine, 1-pyrrolidinepropanamine, n-3-aminopropyl pyrrolidine, 3-pyrrolidinopropylamine, 3-1-pyrrolidinyl propylamine, pyrrolidine, 1-3-aminopropyl, 3-pyrrolidin-1-ylpropylamine, 3-1-pyrrolidino propylamineShow MoreShow Less
PubChem CID 31670
IUPAC Name 3-pyrrolidin-1-ylpropan-1-amine
SMILES C1CCN(C1)CCCN

Specifications

Density 1.014
Boiling Point 191°C to 193°C
Refractive Index 1.471
Quantity 5 g
UN Number UN2735
Beilstein 1191
Sensitivity Air Sensitive
Solubility Information Soluble in water,soluble in chloroform
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material 1-(3-Aminopropyl)pyrrolidine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451676 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberL04739.14,L04739.06
Lot NumberUSLF003676 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL04739.14,L04739.06
Lot Number10232007 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL04739.14,L04739.06
Lot Number10152453 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04739.14,L04739.06
Lot Number10199851 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04739.14,L04739.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(3-Aminopropyl)pyrrolidine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.