missing translation for 'onlineSavingsMsg'
Learn More
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 97%, Thermo Scientific™ Product Code.: 10361583

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 97%, Thermo Scientific™

Product Code. 10361583
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10361583

Brand: Thermo Scientific Maybridge CC33939CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852227-96-4
Molecular Formula C17H26BNO2
Molecular Weight (g/mol) 287.21
MDL Number MFCD07368524
InChI Key OTOKWHGMHAAFRM-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 4-1-piperidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-piperidine, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, amtb222, 4-piperidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl piperidineShow MoreShow Less
PubChem CID 4961250
IUPAC Name 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 287.21
Percent Purity 97%
Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.