missing translation for 'onlineSavingsMsg'
Learn More
1-Adamantanamine, 98%, Thermo Scientific Chemicals Product Code.: 15403867

1-Adamantanamine, 98%, Thermo Scientific Chemicals

Product Code. 15403867
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15403867

Brand: Thermo Scientific Alfa Aesar H30076.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-94-5
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.253
MDL Number MFCD00074732
InChI Key DKNWSYNQZKUICI-UHFFFAOYSA-N
Synonym amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrelShow MoreShow Less
PubChem CID 2130
ChEBI CHEBI:2618
IUPAC Name adamantan-1-amine
SMILES C1C2CC3CC1CC(C2)(C3)N

Specifications

Melting Point ∼200°C (sublimation)
Quantity 5 g
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 151.25
Percent Purity 98%
Chemical Name or Material 1-Adamantanamine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245295 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog NumberH30076.06,H30076.14,H30076.22
Lot Number10177269 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH30076.06,H30076.14,H30076.22
Lot Number10208854 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH30076.06,H30076.14,H30076.22
Lot Number10168547 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH30076.06,H30076.14,H30076.22
Lot Number10177132 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH30076.06,H30076.14,H30076.22
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Adamantanamine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.