missing translation for 'onlineSavingsMsg'
Learn More
1-Methylimidazole, 99%, Thermo Scientific Chemicals Product Code.: 10078531

1-Methylimidazole, 99%, Thermo Scientific Chemicals

Product Code. 10078531
5 kg, Plastic drum
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10078531

Brand: Thermo Scientific Acros 126990050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-47-7
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005292
InChI Key MCTWTZJPVLRJOU-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazoleShow MoreShow Less
PubChem CID 1390
ChEBI CHEBI:113454
IUPAC Name 1-methylimidazole
SMILES CN1C=CN=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -2.0°C
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 198.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Refractive Index 1.4955 to 1.4975
Quantity 5 kg
Beilstein 23, 46
Fieser 05,447
Specific Gravity 1.03
Solubility Information Solubility in water: 145g/L (25°C). Other solubilities: soluble in organic solvents
Formula Weight 82.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Methylimidazole, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461578 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog Number126990050,126995000,126991000
Lot NumberA0449682 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number126990050,126995000,126991000
Lot NumberA0455082 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number126990050,126995000,126991000
Lot NumberA0394732 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number126990050,126995000,126991000
Lot NumberA0412969 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number126990050,126995000,126991000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Methylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methylimidazole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.