missing translation for 'onlineSavingsMsg'
Learn More
1-Naphthyl phosphate, monosodium salt monohydrate, 98+%, Thermo Scientific Chemicals Product Code.: 10124760

1-Naphthyl phosphate, monosodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Product Code. 10124760
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10124760

Brand: Thermo Scientific Acros 226890050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81012-89-7
Molecular Formula C10H7O4P
Molecular Weight (g/mol) 222.14
MDL Number MFCD00150615
InChI Key YNXICDMQCQPQEW-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplcShow MoreShow Less
PubChem CID 45055387
SMILES [O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 189.0°C to 191.0°C
Color White
pH 3.5 to 4.5 (5% aq. soln.)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C10H7OP(O)(OH)ONa·H2O
Quantity 5 g
Solubility Information (5% in water) Clear colorless to light yellow
IUPAC Name naphthalen-1-yl dihydrogen phosphate;sodium;hydrate
Formula Weight 264.15
Percent Purity 98+%
Physical Form Fine Powder
Chemical Name or Material 1-Naphthyl phosphate, monosodium salt monohydrate, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465463 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number226890050,226890010,226891000,226890250
Lot NumberA0453056 Certificate TypeCertificate of Analysis Date11/05/2023 Catalog Number226890050,226890010,226891000,226890250
Lot NumberA0403434 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number226890050,226890010,226891000,226890250
Lot NumberA0402935 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number226890050,226890010,226891000,226890250
Lot NumberA0397394 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number226890050,226890010,226891000,226890250
compliance-icons
Product Identifier
  • 1-Naphthyl phosphate, monosodium salt, monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Naphthyl phosphate, monosodium salt monohydrate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.