missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ 1-step Fix/Lyse Solution (10X)

Enables lysis of RBCs after staining peripheral blood cells with fluorochrome-conjugated antibodies. Formulated to lyse non-nucleated erythrocytes while maintaining fixed and labeled leukocyte population.

Brand:  Invitrogen™ 00-5333-57

228.20 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.


 View more versions of this product

Product Code. 15506296

  • £326.00 / 200mL
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

Contains 10% formaldehyde and 30% diethylene glycol

This item is not returnable. View return policy

Description

Description

  • Isolation of mononuclear cells by gradient centrifugation (e.g. Ficoll-Paque density gradient separation) is not required when using this solution
  • Compatible with antibodies conjugated to organic dyes and tandem dyes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1-step Fixation/Lyse Solution
eBioscience™
200 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2661307 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number00-5333-57
Lot Number2410218 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number00-5333-57
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ eBioscience™ 1-step Fix/Lyse Solution (10X) > 200mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.