Learn More
Thermo Scientific™ Reaction Buffer with MgCl2 for DNase I (10X)
Optimize reaction condions for RNase-free DNase I with this 10X reaction Tris-HCl (pH 7.5) buffer containing MgCl2.
Brand: Thermo Scientific™ B43
11.36 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
10X Reaction Buffer with MgCl2 is used with DNase I, RNase-free.

Specifications
10X | |
1 mL | |
Reaction Buffer | |
Traditional Cloning |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Grow Your Discovery
Fall in love with big savingsPromo Code: 24107Expires: 2025-03-28
Days
1Hours
7Minutes
0Seconds
18 -
Multiple Manufacturers
More Discovery, Less Cost
Now extended - offer ends 28 MarchPromo Code: 24111Expires: 2025-03-28
Days
1Hours
7Minutes
0Seconds
18New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.