Learn More
Bochem™ 18/10 Stainless Steel Lab jack
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Lab jack features improved scissor technology with reduced tolerances.
Brand: Bochem™ 11120
This item is not returnable.
View return policy
Description
- Manufactured from 18/10 stainless steel
- Improved scissor technology with reduced tolerances
- Ergonomic knob, rounded and user friendly
- Scissor-construction made of 18/10-stainless steel
- All models are manufactured according to DIN 12897.
- Maximal dynamic operating load (Kg max. dyn.) and maximal static operating load (Kg max. stat.) norms are according to DIN 12897
- Smaller models (up to 240x240mm; Typ 1) have a simple hand-operated adjusting wheel
- Bigger models (from 300x300mm; Typ 2) recommend the use of included ratchet, which connects with a special adjusting wheel
Specifications
Stainless Steel (18/10) | |
160 mm | |
1 Ea. |
60 to 275 mm | |
Lab Jack |
Product Content Correction
Product Title
Bochem™ 18/10 Stainless Steel Lab jack > Length: 160mm
Spot an opportunity for improvement?Share a Content Correction