Learn More
Fisherbrand™ Aluminum Labjacks
Provides for precise height adjustment with the turn of a screw. Fisherbrand™ Aluminum Labjacks are manufactured to close tolerances with a stainless steel operating screw.
Brand:  Fisherbrand™ 2035
 View more versions of this product
Product Code. 11825091
  • £271.48 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Blue anodized finish
  • Completely stable and free from rocking movements
  • Top and bottom plates are specially designed aluminum extrusion giving exceptional rigid and robust support
Note:
Image shows Labjack with extension plate. (Extension plates are not included; sold separately.)
Specifications
Specifications
Aluminum
150 mm
With stainless steel operating screw
1 Ea.
60 to 290 mm
150 mm
Lab Jack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Aluminum Labjacks > Plate Size: 150 x 150mm; Max Height: 290mm