missing translation for 'onlineSavingsMsg'
Learn More
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals Product Code.: 11451177

2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11451177
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11451177

Brand: Thermo Scientific Alfa Aesar B24161.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.

Solubility
Soluble in water 170 g/L, alcohol, DMSO, ethanol.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 550-99-2
Molecular Formula C14H15ClN2
Molecular Weight (g/mol) 246.738
MDL Number MFCD00012554
InChI Key DJDFFEBSKJCGHC-UHFFFAOYSA-N
Synonym naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon
PubChem CID 11079
ChEBI CHEBI:7470
IUPAC Name 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride
SMILES C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl

Specifications

Melting Point 257°C to 261°C
Quantity 25 g
UN Number UN2811
Beilstein 3716843
Merck Index 14,6368
Solubility Information Soluble in water 170g/L,alcohol,DMSO,ethanol.
Formula Weight 246.74
Percent Purity 99%
Chemical Name or Material 2-(1-Naphthylmethyl)-2-imidazoline hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250944 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB24161.22,B24161.14,B24161.36
Lot Number10248535 Certificate TypeCertificate of Analysis Date24/11/2023 Catalog NumberB24161.22,B24161.14,B24161.36
Lot NumberH4071A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24161.22,B24161.14,B24161.36
Lot Number10228131 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24161.22,B24161.14,B24161.36
Lot Number10230846 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24161.22,B24161.14,B24161.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.