missing translation for 'onlineSavingsMsg'
Learn More
2,2':5',2″-Terthiophene, 99%, Thermo Scientific Chemicals Product Code.: 11412768

2,2':5',2″-Terthiophene, 99%, Thermo Scientific Chemicals

Product Code. 11412768
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11412768

Brand: Thermo Scientific Alfa Aesar A16748.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

2,2?:5?,2??-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′:5′,2′′-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1081-34-1
Molecular Formula C12H8S3
Molecular Weight (g/mol) 248.376
MDL Number MFCD00012167
InChI Key KXSFECAJUBPPFE-UHFFFAOYSA-N
Synonym 2,2':5',2-terthiophene, alpha-terthienyl, alpha-terthiophene, 2,5-di 2-thienyl thiophene, 2,2',5',2-terthienyl, 2,5-bis 2-thienyl thiophene, unii-0p77rau2rr, 2,2'-5',2-terthiophene, 0p77rau2rr, 2,2':5',2-terthienylShow MoreShow Less
PubChem CID 65067
ChEBI CHEBI:10335
IUPAC Name 2,5-dithiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC=C(S2)C3=CC=CS3

Specifications

Melting Point 89°C to 94°C
Boiling Point 160°C (0.1 mmHg)
Quantity 1 g
Beilstein 178604
Merck Index 14,9174
Solubility Information Insoluble in water.
Formula Weight 248.39
Percent Purity 99%
Chemical Name or Material 2,2':5',2''-Terthiophene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI9725A Certificate TypeCertificate of Analysis Date29/07/2022 Catalog NumberA16748.06,A16748.03,A16748.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2':5',2″-Terthiophene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.