missing translation for 'onlineSavingsMsg'
Learn More
2,2'-Bithiophene-5-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals Product Code.: 15487847

2,2'-Bithiophene-5-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Product Code. 15487847
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15487847

Brand: Thermo Scientific Alfa Aesar H37989.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479719-88-5
Molecular Formula C14H17BO2S2
Molecular Weight (g/mol) 292.22
MDL Number MFCD04039974
InChI Key HPOQARMSOPOZMW-UHFFFAOYSA-N
Synonym 2,2'-bithiophene-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,2'-bithiophene, 2-2,2'-bithiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-thiophen-2-yl thiophen-2-yl-1,3,2-dioxaborolane, acmc-209kb7, 4,4,5,5-tetramethyl-2-5-thiophen-2-ylthiophen-2-yl-1,3,2-dioxaborolane, 2,2'-bithiophene-5'-boronic acid pinacol esterShow MoreShow Less
PubChem CID 3592790
IUPAC Name 4,4,5,5-tetramethyl-2-(5-thiophen-2-ylthiophen-2-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1

Specifications

Melting Point 35°C to 38°C
Color Blue-Green to White
Quantity 1 g
Formula Weight 292.2
Percent Purity 98%
Physical Form Fused Solid
Chemical Name or Material 2,2'-Bithiophene-5-boronic acid pinacol ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10240029 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberH37989.06,H37989.03
Lot Number10239582 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberH37989.06,H37989.03
Lot Number10243459 Certificate TypeCertificate of Analysis Date19/10/2022 Catalog NumberH37989.06,H37989.03
Lot Number10243043 Certificate TypeCertificate of Analysis Date31/08/2022 Catalog NumberH37989.06,H37989.03
Lot Number10236376 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH37989.06,H37989.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2'-Bithiophene-5-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.