missing translation for 'onlineSavingsMsg'
Learn More
2,2-Bithiophene-5-carboxylic acid, 97%, Thermo Scientific™ Product Code.: 10674745

2,2-Bithiophene-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10674745
10 g
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy

Product Code. 10674745

Brand: Thermo Scientific Maybridge CC14601EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2060-55-1
Molecular Formula C9H6O2S2
Molecular Weight (g/mol) 210.265
InChI Key PTNWHEHDBNNKEO-UHFFFAOYSA-N
Synonym 2,2'-bithiophene-5-carboxylic acid, 5-thiophen-2-yl thiophene-2-carboxylic acid, 2,2-bithiophene-5-carboxylic acid, 2,2'-bithiophene-5-carboxylicacid, 2,2' bithiophenyl-5-carboxylic acid, 2,2'-bithiophene-5-ca, 5-carboxy-2,2'-bithiophene, 2,2'-bithienyl-5'-carboxylic, 5-2-thienyl-2-thiophenecarboxylic acidShow MoreShow Less
PubChem CID 150965
IUPAC Name 5-thiophen-2-ylthiophene-2-carboxylic acid
SMILES C1=CSC(=C1)C2=CC=C(S2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 210.28
Percent Purity 97%
Chemical Name or Material 2, 2'-Bithiophene-5-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2-Bithiophene-5-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.