missing translation for 'onlineSavingsMsg'
Learn More
2,2'-Bithiophene, 98%, Thermo Scientific Chemicals Product Code.: 11439453

2,2'-Bithiophene, 98%, Thermo Scientific Chemicals

Product Code. 11439453
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11439453

Brand: Thermo Scientific Alfa Aesar A12335.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,2'-Bithiophene is used as a precursor in the preparation of 5,5'-bis(trimethylstannyl)-2,2'-bithiophene , which is used for the synthesis of small molecules and polymer semiconductors for organic field-effect transistor (OFETs), organic light-emitting diode (OLED), Plastic Light Emitting Diode (PLED) and Organic Photovoltaic (OPV) applications. It is associated with aryl iodides and involved in rhodium catalyzed arylation of heteroarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Bithiophene is used as a precursor in the preparation of 5,5′-bis(trimethylstannyl)-2,2′-bithiophene , which is used for the synthesis of small molecules and polymer semiconductors for organic field-effect transistor (OFETs), organic light-emitting diode (OLED), Plastic Light Emitting Diode (PLED) and Organic Photovoltaic (OPV) applications. It is associated with aryl iodides and involved in rhodium catalyzed arylation of heteroarenes.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-97-7
Molecular Formula C8H6S2
Molecular Weight (g/mol) 166.256
MDL Number MFCD00005414
InChI Key OHZAHWOAMVVGEL-UHFFFAOYSA-N
Synonym 2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiopheneShow MoreShow Less
PubChem CID 68120
ChEBI CHEBI:36821
IUPAC Name 2-thiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC=CS2

Specifications

Melting Point 30°C to 33°C
Boiling Point 260°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 3039
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 166.27
Percent Purity 98%
Chemical Name or Material 2,2'-Bithiophene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10190553 Certificate TypeCertificate of Analysis Date20/06/2023 Catalog NumberA12335.14,A12335.03,A12335,A12335.06
Lot Number5002P27S Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12335.03,A12335.14,A12335.06
Lot Number10229368 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12335.03,A12335.14,A12335.06
Lot NumberI6503A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12335.03,A12335.14,A12335.06
Lot Number10135542 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12335.03,A12335.14,A12335.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2'-Bithiophene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.