missing translation for 'onlineSavingsMsg'
Learn More
2,2-Difluoro-1,3-benzodioxole, 97%, Thermo Scientific Chemicals Product Code.: 11418917

2,2-Difluoro-1,3-benzodioxole, 97%, Thermo Scientific Chemicals

Product Code. 11418917
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11418917

Brand: Thermo Scientific Alfa Aesar B20540.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1583-59-1
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00236217
InChI Key DGCOGZQDAXUUBY-UHFFFAOYSA-N
Synonym 2,2-difluorobenzo d 1,3 dioxole, 2,2-difluorobenzodioxole, 2,2-difluoro-2h-1,3-benzodioxole, 1,2-difluoromethylenedioxy benzene, 2,2-difluoro-benzodioxole, 2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 2,2-difluoro, pubchem17570, acmc-1bus1, ksc491c9dShow MoreShow Less
PubChem CID 74103
IUPAC Name 2,2-difluoro-2H-1,3-benzodioxole
SMILES FC1(F)OC2=CC=CC=C2O1

Specifications

Density 1.308
Boiling Point 129°C to 130°C
Flash Point 32°C (90°F)
Refractive Index 1.443
Quantity 25 g
UN Number UN1993
Beilstein 1307648
Formula Weight 158.11
Percent Purity 97%
Chemical Name or Material 2,2-Difluoro-1,3-benzodioxole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10175187 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog NumberB20540.06,B20540.14
Lot Number10170083 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20540.06,B20540.14
Lot Number10138599 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20540.06,B20540.14
Lot Number10167025 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20540.06,B20540.14
Lot Number10175187 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20540.06,B20540.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2-Difluoro-1,3-benzodioxole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.