missing translation for 'onlineSavingsMsg'
Learn More
2-(2-Methyl-1,3-thiazol-4-yl)aniline, 95%, Thermo Scientific™ Product Code.: 10285794

2-(2-Methyl-1,3-thiazol-4-yl)aniline, 95%, Thermo Scientific™

Product Code. 10285794
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10285794

Brand: Thermo Scientific Maybridge CC68414CB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 305811-38-5
Molecular Formula C10H10N2S
Molecular Weight (g/mol) 190.264
MDL Number MFCD09965287
InChI Key AJFBHXXQTQAILP-UHFFFAOYSA-N
Synonym 2-2-methyl-1,3-thiazol-4-yl aniline, 2-2-methyl-4-thiazolyl aniline, 2-2-methylthiazol-4-yl aniline, 2-2-methyl-1,3-thiazol-4-yl phenylamine, 2-2-methyl-1,3-thiazol-4-yl phenyl amineShow MoreShow Less
PubChem CID 22493281
IUPAC Name 2-(2-methyl-1,3-thiazol-4-yl)aniline
SMILES CC1=NC(=CS1)C2=CC=CC=C2N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 190.27
Percent Purity 95%
Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(2-Methyl-1,3-thiazol-4-yl)aniline, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.