missing translation for 'onlineSavingsMsg'
Learn More
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals Product Code.: 10713901

2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals

Product Code. 10713901
25 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10713901

Brand: Thermo Scientific Acros 139250250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1004-38-2
MDL Number MFCD00006100
InChI Key JTTIOYHBNXDJOD-UHFFFAOYSA-N
Synonym 2,4,6-triaminopyrimidine, 2,4,6-pyrimidinetriamine, pyrimidine-2,4,6-triyltriamine, pyrimidine, 2,4,6-triamino, triamino pyrimidine, 3ay, aaat, pubchem7216, 2,6-pyrimidinetriamine, 2,6-triaminopyrimidineShow MoreShow Less
PubChem CID 13863
ChEBI CHEBI:39857
IUPAC Name pyrimidine-2,4,6-triamine
SMILES C1=C(N=C(N=C1N)N)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 246°C to 252°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Molecular Formula C4H7N5
Quantity 25 g
Beilstein 24,47
Solubility Information Solubility in water: 36.5g/kg in water (20°C). Other solubilities: 22.7g/kg in methanol (20°C),4.5g/kg in ethanol (20°C)
Molecular Weight (g/mol) 125.13
Formula Weight 125.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 4, 6-Triaminopyrimidine, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0405113 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number139251000,139250050,139250250
Lot NumberA0444464 Certificate TypeCertificate of Analysis Date09/08/2022 Catalog Number139251000,139250050,139250250
Lot NumberA0442890 Certificate TypeCertificate of Analysis Date09/06/2022 Catalog Number139251000,139250050,139250250
Lot NumberA0405390 Certificate TypeCertificate of Analysis Date18/08/2021 Catalog Number139251000,139250050,139250250
Lot NumberA0419804 Certificate TypeCertificate of Analysis Date29/06/2020 Catalog Number139251000,139250050,139250250
compliance-icons
Product Identifier
  • 2,4,6-Triaminopyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.