missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloropyrido[3,2-d]pyrimidine, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Melting Point | 175.0°C to 177.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Quantity | 250 mg |
Formula Weight | 200.03 |
Percent Purity | 95% |
Chemical Name or Material | 2,4-Dichloropyrido[3,2-d]pyrimidine |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,4-Dichloropyrido[3,2-d]pyrimidine, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction