missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific™ Product Code.: 15470806

2,4-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific™

Product Code. 15470806
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 15470806

Brand: Thermo Scientific Alfa Aesar H54134.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Store
TRUSTED_SUSTAINABILITY

Specifications

CAS 34589-46-3
Molecular Formula C8H9BrMg
MDL Number MFCD01311493
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 2,4-dimethylphenylmagnesium bromide, 2,4-dimethylphenylmagnesium bromide solution, hpgwkalajlxsdh-uhfffaoysa-m, 2,4-dimethylphenylmagne-sium bromide, 2,4-dimethylphenyl magnesium bromide, magnesium,1,3-dimethylbenzene-6-ide,bromide, 2,4-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 2,4-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 2,4-dimethylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information Reacts with water.
InChI Key HPGWKALAJLXSDH-UHFFFAOYSA-M
SMILES CC1=CC=C([Mg]Br)C(C)=C1
IUPAC Name magnesium;1,3-dimethylbenzene-6-ide;bromide
Molecular Weight (g/mol) 209.37
PubChem CID 13637491
Formula Weight 209.37
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 2,4-Dimethylphenylmagnesium bromide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.