missing translation for 'onlineSavingsMsg'
Learn More
2,4-Piperidinedione, 97%, Thermo Scientific Chemicals Product Code.: 11946591

2,4-Piperidinedione, 97%, Thermo Scientific Chemicals

Product Code. 11946591
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11946591

Brand: Thermo Scientific Acros 446730050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50607-30-2
InChI Key RDNZDMDLRIQQAX-UHFFFAOYSA-N
PubChem CID 10887863
IUPAC Name piperidine-2,4-dione
SMILES C1CNC(=O)CC1=O

Specifications

Assay Percent Range 97%
Molecular Formula C5H7NO2
Quantity 5 g
Synonym 2,4-piperidinedione, 2,4-piperadinedione, 2,4-dioxopiperidine, 2,4-diketopiperidine, piperidin-2,4-dione, 2,4 dioxopiperdine, 2, 4-dioxopiperidine, pubchem22543, acmc-209ko8
Molecular Weight (g/mol) 113.116
Formula Weight 113.12
Chemical Name or Material 2, 4-Piperidinedione

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number340283 Certificate TypeCertificate of Analysis Date23/12/2021 Catalog Number446730010,446730050
compliance-icons
Product Identifier
  • 2,4-Piperidinedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Piperidinedione, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.