missing translation for 'onlineSavingsMsg'
Learn More
(-)-Strychnine, 98%, Thermo Scientific Chemicals Product Code.: 10511411

(-)-Strychnine, 98%, Thermo Scientific Chemicals

Product Code. 10511411
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10511411

Brand: Thermo Scientific Acros 158950250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-24-9
Molecular Formula C21H22N2O2
Molecular Weight (g/mol) 334.42
MDL Number MFCD00005941
InChI Key QMGVPVSNSZLJIA-FVWCLLPLSA-N
Synonym strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos
PubChem CID 441071
ChEBI CHEBI:28973
IUPAC Name (4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-oneShow MoreShow Less
SMILES O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 284.0°C to 286.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 8985
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chlororform and hot ethanol,insoluble in diethyl ether
Formula Weight 334.41
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material (-)-Strychnine, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465209 Certificate TypeCertificate of Analysis Date06/08/2024 Catalog Number158951000,158950250
Lot NumberA0375246 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog Number158951000,158950250
Lot NumberA0444044 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number158951000,158950250
Lot NumberA0446429 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog Number158951000,158950250
compliance-icons compliance-icons
Product Identifier
  • (-)-Strychnine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(-)-Strychnine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.