missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dimethoxy-4-methylbenzaldehyde, 97%, Thermo Scientific Chemicals Product Code.: 15471209

2,5-Dimethoxy-4-methylbenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 15471209
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15471209

Brand: Thermo Scientific Alfa Aesar H64140.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the preparation of phenylamine derivative as potential psychotomimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of phenylamine derivative as potential psychotomimetics.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4925-88-6
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD02253192
InChI Key LRSRTWLEJBIAIT-UHFFFAOYSA-N
Synonym 4-methyl-2,5-dimethoxybenzaldehyde, unii-238b3l4auk, 2,5-dimethoxy-4-methyl-benzaldehyde, benzaldehyde, 2,5-dimethoxy-4-methyl, 2,5-dimethoxy-p-tolualdehyde, p-tolualdehyde, 2,5-dimethoxy, who-dd, 2,5-dimethoxy-4-methylbenzaldehyde;, 2,5-dimethoxy-4-methyl-benzaldehyde;, #Show MoreShow Less
PubChem CID 602019
IUPAC Name 2,5-dimethoxy-4-methylbenzaldehyde
SMILES COC1=CC(C)=C(OC)C=C1C=O

Specifications

Melting Point 82°C to 86°C
Color White
Quantity 5 g
Beilstein 1866592
Solubility Information Soluble in dichloromethane,dimethyl sulfoxide and methanol. Slightly soluble in water.
Formula Weight 180.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Dimethoxy-4-methylbenzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU05C003 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH64140.03,H64140.14,H64140.06
Lot NumberS29G025 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH64140.03,H64140.14,H64140.06
Lot NumberY28E065 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH64140.03,H64140.14,H64140.06
Lot Number81303176 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH64140.03,H64140.14,H64140.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dimethoxy-4-methylbenzaldehyde, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.