missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzeneboronic acid, 97%, Thermo Scientific™

POA

Chemical Identifiers

CAS 89694-44-0
Molecular Formula C7H8BBrO3
Molecular Weight (g/mol) 230.852
MDL Number MFCD06659859
InChI Key BJQKEDXKQVNQPR-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzene boronic acid, 2-bromo-5-methoxyphenyl boronic acid, 2-bromo-5-methoxy benzeneboronic acid, acmc-209r2f, amtb841, 2-bromo-5-methoxyphenylboranic acid, 2-bromo-5-methoxyphenylboronicacid, boronic acid,b-2-bromo-5-methoxyphenyl
PubChem CID 25266062
IUPAC Name (2-bromo-5-methoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)OC)Br)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15479737
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52972.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15469737
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52972.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

89694-44-0
230.852
BJQKEDXKQVNQPR-UHFFFAOYSA-N
25266062
B(C1=C(C=CC(=C1)OC)Br)(O)O
C7H8BBrO3
MFCD06659859
2-bromo-5-methoxybenzene boronic acid, 2-bromo-5-methoxyphenyl boronic acid, 2-bromo-5-methoxy benzeneboronic acid, acmc-209r2f, amtb841, 2-bromo-5-methoxyphenylboranic acid, 2-bromo-5-methoxyphenylboronicacid, boronic acid,b-2-bromo-5-methoxyphenyl
(2-bromo-5-methoxyphenyl)boronic acid

Specifications

89694-44-0
C7H8BBrO3
2-bromo-5-methoxybenzene boronic acid, 2-bromo-5-methoxyphenyl boronic acid, 2-bromo-5-methoxy benzeneboronic acid, acmc-209r2f, amtb841, 2-bromo-5-methoxyphenylboranic acid, 2-bromo-5-methoxyphenylboronicacid, boronic acid,b-2-bromo-5-methoxyphenyl
B(C1=C(C=CC(=C1)OC)Br)(O)O
230.852
230.85
2-Bromo-5-methoxybenzeneboronic acid
214°C to 216°C
MFCD06659859
BJQKEDXKQVNQPR-UHFFFAOYSA-N
(2-bromo-5-methoxyphenyl)boronic acid
25266062
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications