missing translation for 'onlineSavingsMsg'
Learn More
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals Product Code.: 11498257

2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals

Product Code. 11498257
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11498257

Brand: Thermo Scientific Alfa Aesar A17625.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.

Solubility
Soluble in water.

Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2576-47-8
Molecular Formula C2H7Br2N
Molecular Weight (g/mol) 204.893
MDL Number MFCD00012886
InChI Key WJAXXWSZNSFVNG-UHFFFAOYSA-N
Synonym 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromideShow MoreShow Less
PubChem CID 2774217
IUPAC Name 2-bromoethanamine;hydrobromide
SMILES C(CBr)N.Br

Specifications

Melting Point 172°C to 176°C
Quantity 250 g
Beilstein 3607605
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 204.9
Percent Purity ≥98%
Chemical Name or Material 2-Bromoethylamine hydrobromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251385 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA17625.0B,A17625.30,A17625.18
Lot Number10228709 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA17625.0B,A17625.30,A17625.18
Lot Number10242233 Certificate TypeCertificate of Analysis Date28/07/2022 Catalog NumberA17625.0B,A17625.30,A17625.18
Lot Number10202825 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17625.0B,A17625.30,A17625.18
Lot Number10225640 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17625.0B,A17625.30,A17625.18
compliance-icons
Product Identifier
  • 2-Bromoethylamine hydrobromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Bromoethylamine hydrobromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.