missing translation for 'onlineSavingsMsg'
Learn More
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals Product Code.: 10486900

2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals

Product Code. 10486900
100 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10486900

Brand: Thermo Scientific Acros 106931000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2576-47-8, 107-09-5
Molecular Formula C2H6BrN·HBr
Molecular Weight (g/mol) 204.9
MDL Number MFCD00012886
InChI Key WJAXXWSZNSFVNG-UHFFFAOYSA-N
Synonym 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromideShow MoreShow Less
PubChem CID 2774217
IUPAC Name 2-bromoethanamine;hydrobromide
SMILES C(CBr)N.Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 172.0°C to 176.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula BrCH2CH2NH2·HBr
Quantity 100 g
Beilstein 04,134
Solubility Information Solubility in water: >500g/L (20°C)
Formula Weight 204.9
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Bromoethylamine hydrobromide, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467310 Certificate TypeCertificate of Analysis Date06/11/2024 Catalog Number106935000,106930250,106932500,106930010,106931000,106930025
Lot NumberA0467039 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number106935000,106930250,106932500,106930010,106931000,106930025
Lot NumberA0463823 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number106935000,106930250,106932500,106930010,106931000,106930025
Lot NumberA0462291 Certificate TypeCertificate of Analysis Date25/04/2024 Catalog Number106935000,106930250,106932500,106930010,106931000,106930025
Lot NumberA0429187 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number106935000,106930250,106932500,106930010,106931000,106930025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.