missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-1,3,2-dioxaphospholane-2-oxide, 95%, Thermo Scientific Chemicals Product Code.: 10286011

2-Chloro-1,3,2-dioxaphospholane-2-oxide, 95%, Thermo Scientific Chemicals

Product Code. 10286011
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10286011

Brand: Thermo Scientific Acros 332060050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6609-64-9
Molecular Formula C2H4ClO3P
Molecular Weight (g/mol) 142.48
MDL Number MFCD00043138
InChI Key SBMUNILHNJLMBF-UHFFFAOYSA-N
Synonym 2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphateShow MoreShow Less
PubChem CID 81087
IUPAC Name 2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide
SMILES C1COP(=O)(O1)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 11.0°C to 14.0°C
Color White
Density 1.5500g/mL
Boiling Point 98.0°C to 99.0°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4540
Quantity 5 g
Specific Gravity 1.55
Formula Weight 142.48
Percent Purity 95%
Physical Form Solid Film
Chemical Name or Material 2-Chloro-1, 3, 2-dioxaphospholane-2-oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462204 Certificate TypeCertificate of Analysis Date04/07/2024 Catalog Number332060250,332061000,332060050
Lot NumberA0455460 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number332060250,332061000,332060050
Lot NumberA0442018 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog Number332060250,332061000,332060050
Lot NumberA0390961 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number332060250,332061000,332060050
Lot NumberA0396676 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number332060250,332061000,332060050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-1,3,2-dioxaphospholane-2-oxide, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.