missing translation for 'onlineSavingsMsg'
Learn More
2'-Deoxyinosine-5'-monophosphate disodium salt, 99%, Thermo Scientific Chemicals Product Code.: 15450357

2'-Deoxyinosine-5'-monophosphate disodium salt, 99%, Thermo Scientific Chemicals

Product Code. 15450357
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15450357

Brand: Thermo Scientific Alfa Aesar J64271.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14999-52-1
Molecular Formula C10H11N4Na2O7P
Molecular Weight (g/mol) 376.17
MDL Number MFCD00134874
InChI Key VDPLPYMIJCLJEF-DZVPOJRENA-L
Synonym sodium 2r,3s,5r-3-hydroxy-5-6-oxo-1h-purin-9 6h-yl tetrahydrofuran-2-yl methyl phosphate, 2'-deoxyinosine-5'-monophosphate, disodium salt, 2'-deoxyinosine 5'-monophosphate disodium salt, 2'-deoxyinosine-5'-monophosphate disodium salt, disodium dimp, 2'-deoxyinosine-5'-phosphate sodium, disodium 2'-deoxyinosine 5'-phosphate, sodium 2r,3s,5r-3-hydroxy-5-6-hydroxy-9h-purin-9-yl tetrahydrofuran-2-yl methyl phosphateShow MoreShow Less
PubChem CID 56776971
SMILES [Na+].[Na+].O[C@H]1C[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=NC2=C1N=CNC2=O

Specifications

Color White
Quantity 1 g
IUPAC Name disodium;[(2R,3S,5R)-3-hydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl phosphate
Formula Weight 376.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Deoxyinosine-5'-monophosphate disodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM06L008 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberJ64271.03
Lot NumberY30J029 Certificate TypeCertificate of Analysis Date28/12/2023 Catalog NumberJ64271.03
Lot NumberS28J043 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog NumberJ64271.03,J64271.06
Lot NumberM03J006 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog NumberJ64271.03
Lot NumberH28Y021 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ64271.03
compliance-icons
Product Identifier
  • 2'-Deoxyinosine-5'-monophosphate disodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2'-Deoxyinosine-5'-monophosphate disodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2'-Deoxyinosine-5'-monophosphate disodium salt, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.