missing translation for 'onlineSavingsMsg'
Learn More
2-Ethyl-5,5-dimethyl-1,3-dioxane solution in acetonitrile (1000mg/L) Product Code.: 15468045

2-Ethyl-5,5-dimethyl-1,3-dioxane solution in acetonitrile (1000mg/L)

Product Code. 15468045
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 15468045

Brand: Thermo Scientific Alfa Aesar H32308.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 768-58-1 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 768-58-1
Molecular Formula C8H16O2
MDL Number MFCD16622268
Quantity 100 mL
UN Number UN1648
Synonym solution in acetonitrile 1000mg/l, 1,3-dioxane, 2-ethyl-5,5-dimethyl
InChI Key QSHOOPIYPOINNH-UHFFFAOYSA-N
SMILES CCC1OCC(CO1)(C)C
IUPAC Name 2-ethyl-5,5-dimethyl-1,3-dioxane
Molecular Weight (g/mol) 144.214
PubChem CID 69852
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material 2-Ethyl-5,5-dimethyl-1,3-dioxane solution in acetonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10210382 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog NumberH32308.AE
Lot Number10155591 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32308.AE
Lot Number10210382 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32308.AE
Lot Number10163160 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32308.AE
compliance-icons compliance-icons
Product Identifier
  • 2-Ethyl-5,5-dimethyl-1,3-dioxane solution in acetonitrile (1000mg/L)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Acetonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Ethyl-5,5-dimethyl-1,3-dioxane solution in acetonitrile (1000mg/L) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.