missing translation for 'onlineSavingsMsg'
Learn More
2-Iodoacetamide, 98%, stab. with ca 5-8% water, Thermo Scientific Chemicals Product Code.: 11467736

2-Iodoacetamide, 98%, stab. with ca 5-8% water, Thermo Scientific Chemicals

Product Code. 11467736
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11467736

Brand: Thermo Scientific Alfa Aesar A14715.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.

Solubility
Soluble in water, dimethyl formamide and ethanol. Slightly soluble in methanol.

Notes
Store in a cool place. Light and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-48-9
Molecular Formula C2H4INO
Molecular Weight (g/mol) 184.964
MDL Number MFCD00008028
InChI Key PGLTVOMIXTUURA-UHFFFAOYSA-N
Synonym iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acidShow MoreShow Less
PubChem CID 3727
IUPAC Name 2-iodoacetamide
SMILES C(C(=O)N)I

Specifications

Melting Point 92°C to 95°(dried)
Assay Percent Range 98%, stabilized with ca 5% water
Quantity 5 g
UN Number UN2811
Beilstein 1739080
Sensitivity Light sensitive
Solubility Information Soluble in water,dimethyl formamide and ethanol. Slightly soluble in methanol.
Formula Weight 184.96
Percent Purity 98%
Chemical Name or Material Iodoacetamide, stabilized with ≈5 to 8% water

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10238354 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14715.22,A14715.14,A14715.06
Lot Number10175201 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14715.22,A14715.14,A14715.06
Lot NumberH5849A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14715.22,A14715.14,A14715.06
Lot Number10217806 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14715.22,A14715.14,A14715.06
Lot NumberA05R045 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14715.22,A14715.14,A14715.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Iodoacetamide, 98%, stab. with ca 5-8% water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.