missing translation for 'onlineSavingsMsg'
Learn More
2-Methyl-1H-indol-5-amine, 97% Product Code.: 10597893

2-Methyl-1H-indol-5-amine, 97%

Product Code. 10597893
Amber Glass Bottle; 1g
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy

Product Code. 10597893

Brand: Thermo Scientific Maybridge AC34875DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7570-49-2
CAS Min % 94.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H10N2
MDL Number MFCD02093426
Quantity 1 g
Synonym 5-amino-2-methylindole, 2-methyl-5-aminoindole, 2-methylindole-5-ylamine, pubchem7273, acmc-209ozq, 2-methylindole-5-amine, 2-methyl-5-amino-1h-indole, bidd:gt0193, 5-amino-2-methyl-1h-indole, 2-methyl-1h-indol-5-ylamine
InChI Key JQULCCZIXYRBSE-UHFFFAOYSA-N
SMILES CC1=CC2=CC(N)=CC=C2N1
IUPAC Name 2-methyl-1H-indol-5-amine
Molecular Weight (g/mol) 146.19
PubChem CID 2733992
Formula Weight 146.19
Percent Purity ≥94%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-Methyl-1H-indol-5-amine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-1H-indol-5-amine, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.